Learn More
Cayman Chemical THUNDERHumn IL6 TRFRET B 96Wel
SDP
Catalog No. 502895352
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-6 in cell culture supernatants
Catalog No. 50-289-5352 Supplier Cayman Chemical Supplier No. 50023296 WELL
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-6 in cell culture supernatants