Learn More
Cayman Chemical THUNDERPhospho-AKT pnT30 96Wel
SDP
Catalog No. 502895263
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-AKT 1/2/3 (T308) in cell lysates
Catalog No. 50-289-5263 Supplier Cayman Chemical Supplier No. 50021096 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-AKT 1/2/3 (T308) in cell lysates