Learn More
Cayman Chemical THUNDERPhospho-BAd112 T 96Wel
SDP
Catalog No. 502896837
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-BAD (S112) in cell lysates
Catalog No. 50-289-6837 Supplier Cayman Chemical Supplier No. 50022796 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-BAD (S112) in cell lysates