Learn More
Cayman Chemical THUNDERPhospho-BTKY223 T 96Wel
SDP
Catalog No. 502892742
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-BTK (Y223) in cell lysates
Catalog No. 50-289-2742 Supplier Cayman Chemical Supplier No. 50024096 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-BTK (Y223) in cell lysates