Learn More
Cayman Chemical THUNDERPhospho-CREBS133 480Wel
SDP
Catalog No. 502893156
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-CREB (S133) and Total CREB in cell lysates
Catalog No. 50-289-3156 Supplier Cayman Chemical Supplier No. 500245480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-CREB (S133) and Total CREB in cell lysates