Learn More
Cayman Chemical THUNDERPhospho-CREBS133 96Wel
SDP
Catalog No. 502896790
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-CREB (S133) in cell lysates
Catalog No. 50-289-6790 Supplier Cayman Chemical Supplier No. 50024396 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-CREB (S133) in cell lysates