Learn More
Cayman Chemical THUNDERPhospho-EGFRY1068 96Wel
SDP
Catalog No. 502892741
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-EGFR (Y1068) in cell lysates
Catalog No. 50-289-2741 Supplier Cayman Chemical Supplier No. 50021896 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-EGFR (Y1068) in cell lysates