Learn More
Cayman Chemical THUNDERPhospho-eIF4ES209 96Wel
SDP
Catalog No. 502893587
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-eIF4E (S209) in cell lysates
Catalog No. 50-289-3587 Supplier Cayman Chemical Supplier No. 50024696 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-eIF4E (S209) in cell lysates