Learn More
Cayman Chemical THUNDERPhospho-GSK3S9 T 96Wel
SDP
Catalog No. 502896964
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-GSK3β (S9) in cell lysates
Catalog No. 50-289-6964 Supplier Cayman Chemical Supplier No. 50025196 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-GSK3β (S9) in cell lysates