Learn More
Cayman Chemical THUNDERPhospho-JNK123T1 96Wel
SDP
Catalog No. 502893934
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-JNK1/2/3 (T183/Y185) in cell lysates
Catalog No. 50-289-3934 Supplier Cayman Chemical Supplier No. 50025496 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-JNK1/2/3 (T183/Y185) in cell lysates