Learn More
Cayman Chemical THUNDERPhospho-LCKY394 T 96Wel
SDP
Catalog No. 502893381
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-LCK (Y394) in cell lysates
Catalog No. 50-289-3381 Supplier Cayman Chemical Supplier No. 50025796 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-LCK (Y394) in cell lysates