Learn More
Cayman Chemical THUNDERPhospho-MEK1S218S 96Wel
SDP
Catalog No. 502895436
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-MEK1 (S218/S222) in cell lysates
Catalog No. 50-289-5436 Supplier Cayman Chemical Supplier No. 50025896 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-MEK1 (S218/S222) in cell lysates