Learn More
Cayman Chemical THUNDERPhospho-NF-kBS536 96Wel
SDP
Catalog No. 502895437
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-NF-κB (S536) in cell lysates
Catalog No. 50-289-5437 Supplier Cayman Chemical Supplier No. 50026596 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-NF-κB (S536) in cell lysates