Learn More
Cayman Chemical THUNDERPhospho-RPS6S235S 96Wel
SDP
Catalog No. 502895813
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Ribosomal Protein S6 (S235/S236) in cell lysates.
Catalog No. 50-289-5813 Supplier Cayman Chemical Supplier No. 50027796 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Ribosomal Protein S6 (S235/S236) in cell lysates.