Learn More
Cayman Chemical THUNDERPhospho-RPS6S240S 96Wel
SDP
Catalog No. 502892425
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Ribosomal Protein S6 (S240/S244) in cell lysates.
Catalog No. 50-289-2425 Supplier Cayman Chemical Supplier No. 50027896 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Ribosomal Protein S6 (S240/S244) in cell lysates.