Learn More
Cayman Chemical THUNDERPhospho-SMAD2S465 96Wel
SDP
Catalog No. 502894124
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SMAD2 (S465/S467) in cell lysates.
Catalog No. 50-289-4124 Supplier Cayman Chemical Supplier No. 50028396 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SMAD2 (S465/S467) in cell lysates.