Learn More
Cayman Chemical THUNDERPhospho-SRCY419 T 96Wel
SDP
Catalog No. 502892589
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SRC (Y419) in cell lysates
Catalog No. 50-289-2589 Supplier Cayman Chemical Supplier No. 50028496 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SRC (Y419) in cell lysates