Learn More
Cayman Chemical THUNDERPhospho-STAT1Y701 96Wel
SDP
Catalog No. 502895277
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT1 (Y701) in cell lysates.
Catalog No. 50-289-5277 Supplier Cayman Chemical Supplier No. 50028596 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT1 (Y701) in cell lysates.