Learn More
Cayman Chemical THUNDERPhospho-STAT4Y693 96Wel
SDP
Catalog No. 502895738
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT4 (Y693) in cell lysates
Catalog No. 50-289-5738 Supplier Cayman Chemical Supplier No. 50029996 WELL
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT4 (Y693) in cell lysates