Learn More
Cayman Chemical THUNDERPhospho-STAT6Y641 96Wel
SDP
Catalog No. 502894120
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT6 (Y641) in cell lysates
Catalog No. 50-289-4120 Supplier Cayman Chemical Supplier No. 50028696 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT6 (Y641) in cell lysates