Learn More
Cayman Chemical THUNDERPhospho4EBP1T37T 480
SDP
Catalog No. 502892454
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-4EBP1 (T37/T46) in cell lysates
Catalog No. 50-289-2454 Supplier Cayman Chemical Supplier No. 500224480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-4EBP1 (T37/T46) in cell lysates