Learn More
Cayman Chemical THUNDERPhosphoBAd112 T 4800
SDP
Catalog No. 502895163
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-BAD (S112) in cell lysates
Catalog No. 50-289-5163 Supplier Cayman Chemical Supplier No. 5002274800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-BAD (S112) in cell lysates