Learn More
Cayman Chemical THUNDERPhosphocMetY1234 480Wel
SDP
Catalog No. 502895485
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Met (Y1234/Y1235) and Total Met in cell lysates
Catalog No. 50-289-5485 Supplier Cayman Chemical Supplier No. 500264480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Met (Y1234/Y1235) and Total Met in cell lysates