Learn More
Cayman Chemical THUNDERPhosphoCREBS133 2400
SDP
Catalog No. 502896577
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-CREB (S133) in cell lysates
Catalog No. 50-289-6577 Supplier Cayman Chemical Supplier No. 5002432400 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-CREB (S133) in cell lysates