Learn More
Cayman Chemical THUNDERPhosphoCREBS133 4800
SDP
Catalog No. 502896578
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-CREB (S133) in cell lysates
Catalog No. 50-289-6578 Supplier Cayman Chemical Supplier No. 5002434800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-CREB (S133) in cell lysates