Learn More
Cayman Chemical THUNDERPhosphoeIF2aS51 4800
SDP
Catalog No. 502896582
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-EIF2α (S51) in cell lysates
Catalog No. 50-289-6582 Supplier Cayman Chemical Supplier No. 5002034800 WELL
Add to Cart
Add to Cart
Request a Quote
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-EIF2α (S51) in cell lysates