Learn More
Cayman Chemical THUNDERPhosphoERK12T202 480
SDP
Catalog No. 502893871
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-ERK1/2 (T202/Y204) in cell lysates.
Catalog No. 50-289-3871 Supplier Cayman Chemical Supplier No. 500206480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-ERK1/2 (T202/Y204) in cell lysates.