Learn More
Cayman Chemical THUNDERPhosphoGSK3S9 T 4800
SDP
Catalog No. 502896479
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-GSK3β (S9) in cell lysates
Catalog No. 50-289-6479 Supplier Cayman Chemical Supplier No. 5002514800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-GSK3β (S9) in cell lysates