Learn More
Cayman Chemical THUNDERPhosphoJNK123T1 2400
SDP
Catalog No. 502893161
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-JNK1/2/3 (T183/Y185) in cell lysates
Catalog No. 50-289-3161 Supplier Cayman Chemical Supplier No. 5002542400 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-JNK1/2/3 (T183/Y185) in cell lysates