Learn More
Cayman Chemical THUNDERPhosphoJNK123T1 4800
SDP
Catalog No. 502893162
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-JNK1/2/3 (T183/Y185) in cell lysates
Catalog No. 50-289-3162 Supplier Cayman Chemical Supplier No. 5002544800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-JNK1/2/3 (T183/Y185) in cell lysates