Learn More
Cayman Chemical THUNDERPhosphoLCKY394 T 480
SDP
Catalog No. 502893665
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-LCK (Y394) in cell lysates
Catalog No. 50-289-3665 Supplier Cayman Chemical Supplier No. 500257480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-LCK (Y394) in cell lysates