Learn More
Cayman Chemical THUNDERPhosphoNFkBS536 9600Wel
SDP
Catalog No. 502895616
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-NF-κB (S536) in cell lysates
Catalog No. 50-289-5616 Supplier Cayman Chemical Supplier No. 5002659600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-NF-κB (S536) in cell lysates