Learn More
Cayman Chemical THUNDERPhosphop38a MAPK 480
SDP
Catalog No. 502896728
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38αβγ MAPK14/11/12 (T180/Y182) in cell lysates
Catalog No. 50-289-6728 Supplier Cayman Chemical Supplier No. 500269480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38αβγ MAPK14/11/12 (T180/Y182) in cell lysates