Learn More
Cayman Chemical THUNDERPhosphop53S15 TR 480
SDP
Catalog No. 502892546
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p53 (S15) in cell lysates
Catalog No. 50-289-2546 Supplier Cayman Chemical Supplier No. 500271480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p53 (S15) in cell lysates