Learn More
Cayman Chemical THUNDERPhosphoRPS6S235S 480
SDP
Catalog No. 502893145
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Ribosomal Protein S6 (S235/S236) in cell lysates.
Catalog No. 50-289-3145 Supplier Cayman Chemical Supplier No. 500277480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Ribosomal Protein S6 (S235/S236) in cell lysates.