Learn More
Cayman Chemical THUNDERPhosphoSRCY419 T 480
SDP
Catalog No. 502892476
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SRC (Y419) in cell lysates
Catalog No. 50-289-2476 Supplier Cayman Chemical Supplier No. 500284480 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-SRC (Y419) in cell lysates