Learn More
Cayman Chemical THUNDERPhosphoTBK1S172 4800
SDP
Catalog No. 502896214
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-TBK1/NAK (S172) in cell lysates
Catalog No. 50-289-6214 Supplier Cayman Chemical Supplier No. 5002894800 WELL
Add to Cart
Add to Cart
Request a Quote
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-TBK1/NAK (S172) in cell lysates