Learn More
Cayman Chemical THUNDERTotal JNK123 TRFR 96
SDP
Catalog No. 502893160
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total JNK1/2/3 in cell lysates.
Catalog No. 50-289-3160 Supplier Cayman Chemical Supplier No. 50025596 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total JNK1/2/3 in cell lysates.