Learn More
Cayman Chemical THUNDRcAMP TRFRET asy K 4800
SDP
Catalog No. 502893309
Encompass_Preferred
A homogeneous competitive TR-FRET immunoassay for quantitative measurement of cAMP in cell lysates
Catalog No. 50-289-3309 Supplier Cayman Chemical Supplier No. 5002934800 WELL
A homogeneous competitive TR-FRET immunoassay for quantitative measurement of cAMP in cell lysates