Learn More
Cayman Chemical THUNDRcAMP TRFRET asy K 960Wel
SDP
Catalog No. 502896136
Encompass_Preferred
A homogeneous competitive TR-FRET immunoassay for quantitative measurement of cAMP in cell lysates
Catalog No. 50-289-6136 Supplier Cayman Chemical Supplier No. 500293960 WELL
A homogeneous competitive TR-FRET immunoassay for quantitative measurement of cAMP in cell lysates