Learn More
Cayman Chemical THUNDREXreme PhosphoRK1 96Wel
SDP
Catalog No. 502895737
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of low levels of Phospho-ERK1/2 (T202/Y204) in cell lysates
Catalog No. 50-289-5737 Supplier Cayman Chemical Supplier No. 50029196 WELL
A sandwich TR-FRET immunoassay for semi-quantitative detection of low levels of Phospho-ERK1/2 (T202/Y204) in cell lysates