Learn More
Cayman Chemical THUNDRHumn GMCSF TRFRET 2400
SDP
Supplier:  Cayman Chemical 5003072400 WELL
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human GM-CSF in cell culture supernatants
Catalog No. 50-289-5203