Learn More
Cayman Chemical THUNDRHumn GMCSF TRFRET 4800
SDP
Catalog No. 502895227
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human GM-CSF in cell culture supernatants
Catalog No. 50-289-5227 Supplier Cayman Chemical Supplier No. 5003074800 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human GM-CSF in cell culture supernatants