Learn More
Cayman Chemical THUNDRHumn GMCSF TRFRET 480Wel
SDP
Catalog No. 502895180
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human GM-CSF in cell culture supernatants
Catalog No. 50-289-5180 Supplier Cayman Chemical Supplier No. 500307480 WELL
Add to Cart
Add to Cart
A sandwich TR-FRET immunoassay for the quantitative measurement of human GM-CSF in cell culture supernatants