Learn More
Cayman Chemical THUNDRHumn GMCSF TRFRET 9600
SDP
Catalog No. 502896014
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human GM-CSF in cell culture supernatants
Catalog No. 50-289-6014 Supplier Cayman Chemical Supplier No. 5003079600 WELL
Add to Cart
Add to Cart
Request a Quote
A sandwich TR-FRET immunoassay for the quantitative measurement of human GM-CSF in cell culture supernatants