Learn More
Cayman Chemical THUNDRHumn IL10 TRFRET 2400Wel
SDP
Catalog No. 502895475
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants
Catalog No. 50-289-5475 Supplier Cayman Chemical Supplier No. 5003022400 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants