Learn More
Cayman Chemical THUNDRHumn IL10 TRFRET 4800Wel
SDP
Catalog No. 502895948
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants
Catalog No. 50-289-5948 Supplier Cayman Chemical Supplier No. 5003024800 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants