Learn More
Cayman Chemical THUNDRHumn IL10 TRFRET 9600Wel
SDP
Catalog No. 502895476
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants
Catalog No. 50-289-5476 Supplier Cayman Chemical Supplier No. 5003029600 WELL
Add to Cart
Add to Cart
Request a Quote
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-10 (CSIF) in cell culture supernatants