Learn More
Cayman Chemical THUNDRHumn IL17A TRFRET 4800
SDP
Catalog No. 502895367
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-17A (IL-17; CTLA-8) in cell culture supernatants
Catalog No. 50-289-5367 Supplier Cayman Chemical Supplier No. 5002924800 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL-17A (IL-17; CTLA-8) in cell culture supernatants