Learn More
Cayman Chemical THUNDRHumn IL18 TRFRET 4800Wel
SDP
Catalog No. 502895587
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL‑18 (IL-1F4) in cell culture supernatants
Catalog No. 50-289-5587 Supplier Cayman Chemical Supplier No. 5003044800 WELL
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL‑18 (IL-1F4) in cell culture supernatants